Search Thermo Fisher Scientific
Type | Spectrometer |
Depth (Metric) | 44 cm |
Description | iXR Raman Spectrometer |
Electrical Requirements | 100/240 V, 47/63 Hz |
Electronic Records and Signatures | CE. UL/CSA/ETL, 21 CRF1040.10 |
For Use With (Application) | Nanotechnology, Material science, Academic research, Pharmaceutical, Geology, Life Science/Bio-materials, Polymer, Automotive/Aero and Cosmetics |
Height (Metric) | 37 cm |
Laser Power | Absolute excitation laser power at the sample controlled by OMNIC software |
Model | iXR |
Operating Temperature Range | 16°C to 27 °C |
Resolution | Full range grating: average 2 cm-1/CCD pixel element High resolution grating: average 1 cm-1/CCD pixel element |
Software Details | Thermo Scientific OMNIC software suite |
Weight (Metric) | 25 kg |
Width (Metric) | 29 cm |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
IQLAADGABFFAHCMBJH | Each | iXR Raman Spectrometer | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|